missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 4-[4-(hydroxymethyl)benzyl]tetrahydro-1(2H)-pyrazinecarboxylate, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS | 622381-67-3 |
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Molecular Formula | C17H26N2O3 |
MDL Number | MFCD06658975 |
Quantity | 1 g |
Synonym | tert-butyl 4-4-hydroxymethyl benzyl tetrahydro-1 2h-pyrazinecarboxylate, tert-butyl 4-4-hydroxymethyl benzyl piperazine-1-carboxylate, tert-butyl 4-4-hydroxymethyl phenyl methyl piperazine-1-carboxylate, 1-piperazinecarboxylicacid, 4-4-hydroxymethyl phenyl methyl-, 1,1-dimethylethyl ester, tert-butyl 4-4-hydroxymethyl benzyl tetrahydro-1 2h-pyrazine carboxylate |
InChI Key | KLDBKWMXUMLYJO-UHFFFAOYSA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'