missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 4-[4-(hydroxymethyl)-1,3-thiazol-2-yl]piperidine-1-carboxylate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 95% |
Quantity | 250 mg |
Formula Weight | 298.4 |
Percent Purity | 95% |
Chemical Name or Material | tert-Butyl 4-[4-(hydroxymethyl)-1, 3-thiazol-2-yl]piperidine-1-carboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'