missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Maybridge tert-Butyl 4-(4-formylphenyl)tetrahydro-1(2H)-pyrazinecarboxylate, 1g, 97%

Brand:  Thermo Scientific Maybridge CC30004DA

tert-Butyl 4-(4-formylphenyl)tetrahydro-1(2H)-pyrazinecarboxylate, 1g, 97%

Code : 31

Additional Details : Weight : 0.05100kg

Product Code. 10404193

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.