missing translation for 'onlineSavingsMsg'
Learn More

tert-butyl 4-[4-(aminomethyl)benzyloxy]piperidine-1-carboxylate, 90%, Thermo Scientific™

Catalog Number p-3880865
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
946409-35-4
C18H28N2O3
320.433
MFCD12198114
NZWAUTDCSSMVGG-UHFFFAOYSA-N
tert-butyl 4-4-aminomethyl benzyloxy piperidine-1-carboxylate, tert-butyl 4-4-aminomethyl phenyl methoxy piperidine-1-carboxylate
43811047
tert-butyl 4-[[4-(aminomethyl)phenyl]methoxy]piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)OCC2=CC=C(C=C2)CN
This item is not returnable. View return policy
946409-35-4
C18H28N2O3
320.433
MFCD12198114
NZWAUTDCSSMVGG-UHFFFAOYSA-N
tert-butyl 4-4-aminomethyl benzyloxy piperidine-1-carboxylate, tert-butyl 4-4-aminomethyl phenyl methoxy piperidine-1-carboxylate
43811047
tert-butyl 4-[[4-(aminomethyl)phenyl]methoxy]piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)OCC2=CC=C(C=C2)CN
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 320.43
Percent Purity 90%
Chemical Name or Material Tert-butyl 4-[4-(aminomethyl)benzyloxy]piperidine-1-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'