missing translation for 'onlineSavingsMsg'
Learn More

tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate, 97%, Maybridge™

Catalog Number p-3880951
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1048970-17-7
C16H30BNO4
311.23
MFCD09997792
IBLQMWKHENBVJE-UHFFFAOYSA-N
tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl piperidine-1-carboxylate, n-boc-piperidine-4-boronic acid pinacol ester, 1-boc-piperidin-4-ylboronic acid pinacol ester, 1-boc-piperidine-4-boronic acid pinacol ester, tert-butyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl piperidine-1-carboxylate, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-piperidine-1-carboxylate, 1-boc-piperidin-4-ylboronic acid, pinacol ester, tert-butoxycarbonylpiperidine-4-boronic acid pinacol ester, 1-tert-butoxycarbonyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl piperidine, pubchem21000
43811073
tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
1048970-17-7
C16H30BNO4
311.23
MFCD09997792
IBLQMWKHENBVJE-UHFFFAOYSA-N
tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl piperidine-1-carboxylate, n-boc-piperidine-4-boronic acid pinacol ester, 1-boc-piperidin-4-ylboronic acid pinacol ester, 1-boc-piperidine-4-boronic acid pinacol ester, tert-butyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl piperidine-1-carboxylate, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-piperidine-1-carboxylate, 1-boc-piperidin-4-ylboronic acid, pinacol ester, tert-butoxycarbonylpiperidine-4-boronic acid pinacol ester, 1-tert-butoxycarbonyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl piperidine, pubchem21000
43811073
tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)B1OC(C)(C)C(C)(C)O1
CAS Min % 97.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 311.23
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'