missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 4-(3-hydroxypropyl)tetrahydro-1(2H)-pyridinecarboxylate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 243.34 |
Percent Purity | 95% |
Chemical Name or Material | tert-Butyl 4-(3-hydroxypropyl)tetrahydro-1(2H)-pyridinecarboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'