Learn More
Thermo Scientific Maybridge tert-Butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate, 1g, 97%
Brand: Thermo Scientific Maybridge AW00470DA
tert-Butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate, 1g, 97%
Code : 31
Additional Details : CAS Number : 77279-24-4 Weight : 0.05100kg Transport : UN number : 3259 Chem class : 8 Pack group : III