missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(2-formylphenyl)piperazine-1-carboxylate, 97%, Thermo Scientific™

Catalog Number p-3111123
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
174855-57-3
C16H22N2O3
290.363
MFCD05864664
FGJACYJASSSXNJ-UHFFFAOYSA-N
1-boc-4-2-formylphenyl piperazine, tert-butyl 4-2-formylphenyl piperazine-1-carboxylate, 4-2-formylphenyl piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 4-2-formylphenyl piperazinecarboxylate, 4-2-formyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, acmc-20a0zo, n-boc-4-2-formylphenyl-piperazine, 1-n-boc-4-2-formylphenyl piperazine, 1-boc-4-2-formyl-phenyl-piperazine
6490990
tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2C=O
This item is not returnable. View return policy
174855-57-3
C16H22N2O3
290.363
MFCD05864664
FGJACYJASSSXNJ-UHFFFAOYSA-N
1-boc-4-2-formylphenyl piperazine, tert-butyl 4-2-formylphenyl piperazine-1-carboxylate, 4-2-formylphenyl piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 4-2-formylphenyl piperazinecarboxylate, 4-2-formyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, acmc-20a0zo, n-boc-4-2-formylphenyl-piperazine, 1-n-boc-4-2-formylphenyl piperazine, 1-boc-4-2-formyl-phenyl-piperazine
6490990
tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2C=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 290.36
Chemical Name or Material Tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'