missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (3S)-3-aminobutanoate, 95%, Thermo Scientific Chemicals

Catalog Number 10595223
Click to view available options
Quantity:
100 mg
250 mg
1 g
This item is not returnable. View return policy
161105-54-0
MFCD00798294
BFFNZGWJTHWUMY-LURJTMIESA-N
7021083
tert-butyl (3S)-3-aminobutanoate
CC(CC(=O)OC(C)(C)C)N
This item is not returnable. View return policy
161105-54-0
MFCD00798294
BFFNZGWJTHWUMY-LURJTMIESA-N
7021083
tert-butyl (3S)-3-aminobutanoate
CC(CC(=O)OC(C)(C)C)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Assay Percent Range 94% min. (GC)
Molecular Formula C8H17NO2
Infrared Spectrum Authentic
Packaging Glass Bottle
Synonym tert-butyl 3s-3-aminobutanoate, s-tert-butyl 3-aminobutanoate, butanoic acid, 3-amino-, 1,1-dimethylethyl ester, 3s, tert-butyl-3s-3-aminobutanoate, tert-butyl s-3-aminobutyrate, 1,1-dimethylethyl 35-3-aminobutanoate, s-3-aminobutyric acid tert-butyl ester, 1,1-dimethylethyl 3s-3-aminobutanoate, butanoic acid,3-amino-, 1,1-dimethylethyl ester, 3s
Refractive Index 1.4230 to 1.427
Specific Rotation + 24
Molecular Weight (g/mol) 159.23
Formula Weight 159.23
Percent Purity 95%
Specific Rotation Condition + 24.00 (20.00°C c=1,CHCl3)
Quantity 250 mg
Physical Form Liquid
Chemical Name or Material tert-Butyl (3S)-3-aminobutanoate, 95%
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butyl (3S)-3-aminobutanoate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'