missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl (3R)-3-amino-3-phenylpropanoate, 97%, ACROS Organics™
£99.00 - £362.93
Chemical Identifiers
CAS | 161671-34-7 |
---|---|
Molecular Formula | C13H19NO2 |
Molecular Weight (g/mol) | 221.3 |
MDL Number | MFCD00798309 |
InChI Key | TYYCBAISLMKLMT-LLVKDONJSA-N |
Synonym | tert-butyl 3r-3-amino-3-phenylpropanoate, r-tert-butyl 3-amino-3-phenylpropanoate, 3r-3-phenyl-beta-alanine tert-butyl ester, 1,1-dimethylethyl 3r-3-amino-3-phenylpropanoate, tert-butyl-3r-3-amino-3-phenylpropanoate, benzenepropanoic acid,b-amino-,1,1-dimethylethyl ester, br, benzenepropanoic acid, b-amino-, 1,1-dimethylethyl ester, br |
PubChem CID | 2733824 |
IUPAC Name | tert-butyl (3R)-3-amino-3-phenylpropanoate |
SMILES | CC(C)(C)OC(=O)CC(C1=CC=CC=C1)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10057592
|
Acros Organics
337882500 |
250mg |
£99.00
250mg |
Estimated Shipment 06-05-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10464415
|
Acros Organics
337880010 |
1g |
£362.93
1g |
Estimated Shipment 06-05-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
161671-34-7 | |
221.3 | |
TYYCBAISLMKLMT-LLVKDONJSA-N | |
2733824 | |
CC(C)(C)OC(=O)CC(C1=CC=CC=C1)N |
C13H19NO2 | |
MFCD00798309 | |
tert-butyl 3r-3-amino-3-phenylpropanoate, r-tert-butyl 3-amino-3-phenylpropanoate, 3r-3-phenyl-beta-alanine tert-butyl ester, 1,1-dimethylethyl 3r-3-amino-3-phenylpropanoate, tert-butyl-3r-3-amino-3-phenylpropanoate, benzenepropanoic acid,b-amino-,1,1-dimethylethyl ester, br, benzenepropanoic acid, b-amino-, 1,1-dimethylethyl ester, br | |
tert-butyl (3R)-3-amino-3-phenylpropanoate |
Specifications
tert-Butyl (3R)-3-amino-3-phenylpropanoate | |
Glass Bottle | |
+ 17.00 (20.00°C c=1,CHCl3) | |
Colorless to Yellow | |
97% | |
97% | |
MFCD00798309 | |
TYYCBAISLMKLMT-LLVKDONJSA-N | |
tert-butyl (3R)-3-amino-3-phenylpropanoate | |
2733824 | |
Slightly Viscous Liquid |
Authentic | |
1.4960 to 1.498 | |
+ 17 | |
250mg | |
161671-34-7 | |
C13H19NO2 | |
tert-butyl 3r-3-amino-3-phenylpropanoate, r-tert-butyl 3-amino-3-phenylpropanoate, 3r-3-phenyl-beta-alanine tert-butyl ester, 1,1-dimethylethyl 3r-3-amino-3-phenylpropanoate, tert-butyl-3r-3-amino-3-phenylpropanoate, benzenepropanoic acid,b-amino-,1,1-dimethylethyl ester, br, benzenepropanoic acid, b-amino-, 1,1-dimethylethyl ester, br | |
CC(C)(C)OC(=O)CC(C1=CC=CC=C1)N | |
221.3 | |
221.3 | |
97% |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- tert-Butyl (3R)-3-amino-3-phenylpropanoate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.