missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 3-(thiocarbamoyl)piperidine-1-carboxylate, 95%, Thermo Scientific™

Catalog Number p-3629745
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
274682-80-3
C11H20N2O2S
244.353
MFCD10700060
ZCELPXRDWJCIJE-UHFFFAOYSA-N
tert-butyl 3-thiocarbamoyl piperidine-1-carboxylate, 1-tert-butoxycarbonyl piperidine-3-thiocarboxamide, 3-thiocarbamoyl-piperidine-1-carboxylic acid tert-butyl ester, 1-tert-butoxycarbonyl-3-carbamothioylpiperidine, 3-carbamothioylpiperidine, n1-boc protected, 3-carbamothioylpiperidine,n1-boc protected, tert-butyl3-carbamothioylpiperidine-1-carboxylate, tert-butyl 3-aminothioxomethyl piperidinecarboxylate, tert-butyl 3-aminocarbonothioyl piperidine-1-carboxylate, 1-piperidinecarboxylicacid, 3-aminothioxomethyl-, 1,1-dimethylethyl ester
23546083
tert-butyl 3-carbamothioylpiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)C(=S)N
This item is not returnable. View return policy
274682-80-3
C11H20N2O2S
244.353
MFCD10700060
ZCELPXRDWJCIJE-UHFFFAOYSA-N
tert-butyl 3-thiocarbamoyl piperidine-1-carboxylate, 1-tert-butoxycarbonyl piperidine-3-thiocarboxamide, 3-thiocarbamoyl-piperidine-1-carboxylic acid tert-butyl ester, 1-tert-butoxycarbonyl-3-carbamothioylpiperidine, 3-carbamothioylpiperidine, n1-boc protected, 3-carbamothioylpiperidine,n1-boc protected, tert-butyl3-carbamothioylpiperidine-1-carboxylate, tert-butyl 3-aminothioxomethyl piperidinecarboxylate, tert-butyl 3-aminocarbonothioyl piperidine-1-carboxylate, 1-piperidinecarboxylicacid, 3-aminothioxomethyl-, 1,1-dimethylethyl ester
23546083
tert-butyl 3-carbamothioylpiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)C(=S)N
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 244.36
Percent Purity 95%
Chemical Name or Material Tert-butyl 3-(thiocarbamoyl)piperidine-1-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'