missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 3-bromo-2-furoate, 97%, Thermo Scientific™

Catalog Number p-3629746
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
59862-90-7
C9H11BrO3
247.088
MFCD11109336
STWBDYGNHJTHOY-UHFFFAOYSA-N
tert-butyl 3-bromo-2-furoate, 2-furancarboxylic acid, 3-bromo-, 1,1-dimethylethyl ester
33589622
tert-butyl 3-bromofuran-2-carboxylate
CC(C)(C)OC(=O)C1=C(C=CO1)Br
This item is not returnable. View return policy
59862-90-7
C9H11BrO3
247.088
MFCD11109336
STWBDYGNHJTHOY-UHFFFAOYSA-N
tert-butyl 3-bromo-2-furoate, 2-furancarboxylic acid, 3-bromo-, 1,1-dimethylethyl ester
33589622
tert-butyl 3-bromofuran-2-carboxylate
CC(C)(C)OC(=O)C1=C(C=CO1)Br
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 247.09
Percent Purity 97%
Chemical Name or Material tert-Butyl 3-bromo-2-furoate
compliance-icons
Product Identifier
  • tert-Butyl 3-bromo-2-furoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'