missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-chloro-1,3-thiazole-5-carboxylate, 95%, Thermo Scientific™

Catalog Number p-3586895
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
934570-60-2
C8H10ClNO2S
219.683
MFCD09879991
SQZXVWDARSIXTM-UHFFFAOYSA-N
tert-butyl 2-chlorothiazole-5-carboxylate, t-butyl-2-chloro-1,3-thiazole-5-carboxylate
24229786
tert-butyl 2-chloro-1,3-thiazole-5-carboxylate
CC(C)(C)OC(=O)C1=CN=C(S1)Cl
This item is not returnable. View return policy
934570-60-2
C8H10ClNO2S
219.683
MFCD09879991
SQZXVWDARSIXTM-UHFFFAOYSA-N
tert-butyl 2-chlorothiazole-5-carboxylate, t-butyl-2-chloro-1,3-thiazole-5-carboxylate
24229786
tert-butyl 2-chloro-1,3-thiazole-5-carboxylate
CC(C)(C)OC(=O)C1=CN=C(S1)Cl
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 219.69
Percent Purity 95%
Chemical Name or Material tert-Butyl 2-chloro-1, 3-thiazole-5-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'