missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-amino-2-thioxoethylcarbamate, 90%, Maybridge

£39.00 - £290.00

Chemical Identifiers

CAS 89226-13-1
Molecular Formula C7H14N2O2S
Molecular Weight (g/mol) 190.261
MDL Number MFCD09025922
InChI Key AGBIUUFZUPNDTM-UHFFFAOYSA-N
Synonym tert-butyl 2-amino-2-thioxoethyl carbamate, tert-butyl 2-amino-2-thioxoethylcarbamate, tert-butyl n-carbamothioylmethyl carbamate, tert-butyl n-2-amino-2-sulfanylideneethyl carbamate, carbamic acid, 2-amino-2-thioxoethyl-, 1,1-dimethylethyl ester, carbamic acid,n-2-amino-2-thioxoethyl-, 1,1-dimethylethyl ester, acmc-20ljf8, n-tert-butoxycarbonylglycinethioamide, tert-butoxycarbonylamino acetothioamide, n-t-butoxycarbonylamino acetothioamide
PubChem CID 5324304
IUPAC Name tert-butyl N-(2-amino-2-sulfanylideneethyl)carbamate
SMILES CC(C)(C)OC(=O)NCC(=S)N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10443245 SDS View Documents Maybridge
MO07925DA
1g Amber Glass Bottle £39.00
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 10-06-2020
Please to purchase this item. Need a web account? Register with us today!
10051674 SDS View Documents Maybridge
MO07925EA
10g Amber Glass Bottle £290.00
10g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 08-07-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

89226-13-1
190.261
AGBIUUFZUPNDTM-UHFFFAOYSA-N
5324304
CC(C)(C)OC(=O)NCC(=S)N
C7H14N2O2S
MFCD09025922
tert-butyl 2-amino-2-thioxoethyl carbamate, tert-butyl 2-amino-2-thioxoethylcarbamate, tert-butyl n-carbamothioylmethyl carbamate, tert-butyl n-2-amino-2-sulfanylideneethyl carbamate, carbamic acid, 2-amino-2-thioxoethyl-, 1,1-dimethylethyl ester, carbamic acid,n-2-amino-2-thioxoethyl-, 1,1-dimethylethyl ester, acmc-20ljf8, n-tert-butoxycarbonylglycinethioamide, tert-butoxycarbonylamino acetothioamide, n-t-butoxycarbonylamino acetothioamide
tert-butyl N-(2-amino-2-sulfanylideneethyl)carbamate

Specifications

Tert-butyl 2-amino-2-thioxoethylcarbamate
1g
90.0
C7H14N2O2S
tert-butyl 2-amino-2-thioxoethyl carbamate, tert-butyl 2-amino-2-thioxoethylcarbamate, tert-butyl n-carbamothioylmethyl carbamate, tert-butyl n-2-amino-2-sulfanylideneethyl carbamate, carbamic acid, 2-amino-2-thioxoethyl-, 1,1-dimethylethyl ester, carbamic acid,n-2-amino-2-thioxoethyl-, 1,1-dimethylethyl ester, acmc-20ljf8, n-tert-butoxycarbonylglycinethioamide, tert-butoxycarbonylamino acetothioamide, n-t-butoxycarbonylamino acetothioamide
CC(C)(C)OC(=O)NCC(=S)N
190.261
190.27
Amber Glass Bottle
89226-13-1
90%
MFCD09025922
AGBIUUFZUPNDTM-UHFFFAOYSA-N
tert-butyl N-(2-amino-2-sulfanylideneethyl)carbamate
5324304
90%