missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(tert-Butoxycarbonylmethylene)triphenylphosphorane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
(tert-Butoxycarbonylmethylene)triphenylphosphorane is a chemical used in the synthesis of aldose reductase inhibitors, a podophyllotoxin derivative and tautomycin, a podophyllotoxin derivative and tautomycin. It Is also used in medicinal chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(tert-Butoxycarbonylmethylene)triphenylphosphorane is a chemical used in the synthesis of aldose reductase inhibitors, a podophyllotoxin derivative and tautomycin, a podophyllotoxin derivative and tautomycin. It Is also used in medicinal chemistry.
Solubility
Sparingly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Incompatible with strong oxidizing agents.
(tert-Butoxycarbonylmethylene)triphenylphosphorane is a chemical used in the synthesis of aldose reductase inhibitors, a podophyllotoxin derivative and tautomycin, a podophyllotoxin derivative and tautomycin. It Is also used in medicinal chemistry.
Solubility
Sparingly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 146°C to 150°C |
Linear Formula | (CH3)3CO2CCH=P(C6H5)3 |
Quantity | 25 g |
UN Number | UN2811 |
Beilstein | 2759875 |
Solubility Information | Sparingly soluble in water. |
Formula Weight | 376.44 |
Percent Purity | 98% |
Chemical Name or Material | (tert-Butoxycarbonylmethylene)triphenylphosphorane |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'