missing translation for 'onlineSavingsMsg'
Learn More

(tert-Butoxycarbonylmethylene)triphenylphosphorane, 97%, ACROS Organics™

£18.30 - £194.90

Chemical Identifiers

CAS 35000-38-5
Molecular Formula C24H25O2P
Molecular Weight (g/mol) 376.44
MDL Number MFCD00075545
InChI Key ZWZUFQPXYVYAFO-UHFFFAOYSA-N
Synonym tert-butyl 2-triphenylphosphoranylidene acetate, tert-butoxycarbonylmethylene triphenylphosphorane, tert-butyl triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, 1,1-dimethylethyl ester, t-butoxycarbonylmethylene triphenylphosphorane, tert-butyl 2-triphenyl-??-phosphanylidene acetate, triphenylthoranylidene acetic acid 1,1-dimethylethyl ester, pubchem14371, tert-butyl 2-triphenyl, tert-butyl 2-triphenylylidene acetate
PubChem CID 256127
IUPAC Name tert-butyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
SMILES CC(C)(C)OC(=O)C=P(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10324412 SDS View Documents Acros Organics
370620010
1g Glass bottle £18.30
1g View alternative products
Save Up to 
»
EU Stock 3 Please to purchase this item. Need a web account? Register with us today!
10038921 SDS View Documents Acros Organics
370620050
5g Glass bottle £58.30
5g View alternative products
Save Up to 
»
EU Stock 4 Please to purchase this item. Need a web account? Register with us today!
10048921 SDS View Documents Acros Organics
370620250
25g Glass bottle £194.90
25g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 11-03-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

35000-38-5
376.44
ZWZUFQPXYVYAFO-UHFFFAOYSA-N
256127
CC(C)(C)OC(=O)C=P(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
C24H25O2P
MFCD00075545
tert-butyl 2-triphenylphosphoranylidene acetate, tert-butoxycarbonylmethylene triphenylphosphorane, tert-butyl triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, 1,1-dimethylethyl ester, t-butoxycarbonylmethylene triphenylphosphorane, tert-butyl 2-triphenyl-??-phosphanylidene acetate, triphenylthoranylidene acetic acid 1,1-dimethylethyl ester, pubchem14371, tert-butyl 2-triphenyl, tert-butyl 2-triphenylylidene acetate
tert-butyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate

Specifications

(tert-Butoxycarbonylmethylene)triphenylphosphorane
Glass bottle
146.0°C to 151.0°C
35000-38-5
100.0
C24H25O2P
MFCD00075545
ZWZUFQPXYVYAFO-UHFFFAOYSA-N
tert-butyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
256127
Crystalline Powder
Authentic
White
1g
96.0
97%
(C6H5)3P=CHCO2C(CH3)3
tert-butyl 2-triphenylphosphoranylidene acetate, tert-butoxycarbonylmethylene triphenylphosphorane, tert-butyl triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, 1,1-dimethylethyl ester, t-butoxycarbonylmethylene triphenylphosphorane, tert-butyl 2-triphenyl-??-phosphanylidene acetate, triphenylthoranylidene acetic acid 1,1-dimethylethyl ester, pubchem14371, tert-butyl 2-triphenyl, tert-butyl 2-triphenylylidene acetate
CC(C)(C)OC(=O)C=P(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
376.44
376.44
97%