missing translation for 'onlineSavingsMsg'
Learn More

Tert-Butanol, puriss. p.a. ACS Reagent, ≥99.7% (GC), Honeywell Riedel-de Haën™

Catalog Number p-7120859
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
Packaging:
Poly Bottle
75-65-0
C4H10O
74.123
MFCD00004464
DKGAVHZHDRPRBM-UHFFFAOYSA-N
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
6386
CHEBI:45895
2-methylpropan-2-ol
CC(C)(C)O
75-65-0
C4H10O
74.123
MFCD00004464
DKGAVHZHDRPRBM-UHFFFAOYSA-N
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
6386
CHEBI:45895
2-methylpropan-2-ol
CC(C)(C)O

Puriss. p.a., ACS reagent,≥99.7% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY
Melting Point 25°C
Color Colorless
Linear Formula (CH3)3COH
UN Number UN1120
Beilstein 906698
Formula Weight 74.12g/mol
Percent Purity ≥99.7% (GC)
Grade ACS/puriss. p.a.
Density 0.775g/mL at 25°C
Boiling Point 83°C
Quantity 500 mL
Explosion Limit 0.08
Flash Point 11°C (51.8°F)
Ignition Point 896°F
Packaging Poly Bottle
Physical Form Liquid
Refractive Index n20/D 1.387(lit.)
Vapor Pressure 44mmHg (26°C), 31mmHg (20°C)
Chemical Name or Material Tert-Butanol
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'