missing translation for 'onlineSavingsMsg'
Learn More

α-Terpineol, 97%, mixture of α- and γ-terpineol, ACROS Organics™

Brand:  Acros Organics 203742500

Code : 29

Additional Details : CAS Number : 98-55-5 Weight : 0.30000kg


Quantity 250g
Packaging Glass bottle

 View more versions of this product

Product Code. 10656422

  • £47.80 / 250g


Estimated Shipment
15-06-2018
Add to basket


Specifications

Chemical Identifiers

α-Terpineol
586-81-2
100.0
97%
154.25
p-Menth-1-en-8-ol
97%
96.0
≥96.0%
C10H18O
MFCD00001557

Specifications

α-Terpineol
586-81-2
100.0
0.933g/mL
Liquid
97%
154.25
06, II, 66
p-Menth-1-en-8-ol
1.4820 to 1.4850 (20°C, 589nm)
Clear colorless to slightly yellow
97%
96.0
alpha-terpineol: 85 to 90%, gamma-terpineol: balance (GC)
Authentic
≥96.0%
C10H18O
MFCD00001557
15, 9313
Glass bottle
0.93 to 0.936 (25°/25°C)
250g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • alpha-Terpineol, mixture of alpha- and gamma- terpineol

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS