missing translation for 'onlineSavingsMsg'
Learn More

α-Terpinene, 90%, Tech., Thermo Scientific™

£21.50 - £78.90

Chemical Identifiers

CAS 99-86-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00001534
InChI Key YHQGMYUVUMAZJR-UHFFFAOYSA-N
Synonym alpha-terpinene, p-mentha-1,3-diene, terpilene, 1-isopropyl-4-methyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1-methyl-4-1-methylethyl, alpha-terpinen, 1-methyl-4-1-methylethyl-1,3-cyclohexadiene, 1-methyl-4-isopropylcyclohexadiene-1,3, .alpha.-terpinen, .alpha.-terpinene
PubChem CID 7462
ChEBI CHEBI:10334
IUPAC Name 1-methyl-4-propan-2-ylcyclohexa-1,3-diene
SMILES CC(C)C1=CC=C(C)CC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10561651
SDS View Documents
Thermo Scientific Acros
207491000
100mL Glass bottle
£21.50
100mL
View alternative products
Save Up to 
»
In stock, ready to ship: 8
Please to purchase this item. Need a web account? Register with us today!
10012640
SDS View Documents
Thermo Scientific Acros
207495000
500mL Glass bottle
£78.90
500mL
View alternative products
Save Up to 
»
In stock, ready to ship: 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

99-86-5
136.24
YHQGMYUVUMAZJR-UHFFFAOYSA-N
7462
1-methyl-4-propan-2-ylcyclohexa-1,3-diene
C10H16
MFCD00001534
alpha-terpinene, p-mentha-1,3-diene, terpilene, 1-isopropyl-4-methyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1-methyl-4-1-methylethyl, alpha-terpinen, 1-methyl-4-1-methylethyl-1,3-cyclohexadiene, 1-methyl-4-isopropylcyclohexadiene-1,3, .alpha.-terpinen, .alpha.-terpinene
CHEBI:10334
CC(C)C1=CC=C(C)CC1

Specifications

α-Terpinene
46°C
Glass bottle
0.837
Colorless to Yellow
tech., 90%
88.0
88% min. (GC)
MFCD00001534
14, 9170
Solubility in water: insoluble. Other solubilities: miscible with alcohol,ether
CC(C)C1=CC=C(C)CC1
136.24
CHEBI:10334
Liquid
Technical
0.8370g/mL
Authentic
1.4770 to 1.479
173°C to 175°C
100mL
99-86-5
100.0
C10H16
05, 126
alpha-terpinene, p-mentha-1,3-diene, terpilene, 1-isopropyl-4-methyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1-methyl-4-1-methylethyl, alpha-terpinen, 1-methyl-4-1-methylethyl-1,3-cyclohexadiene, 1-methyl-4-isopropylcyclohexadiene-1,3, .alpha.-terpinen, .alpha.-terpinene
YHQGMYUVUMAZJR-UHFFFAOYSA-N
1-methyl-4-propan-2-ylcyclohexa-1,3-diene
7462
136.24
90%
Safety and Handling

Safety and Handling

Product Identifier
  • alpha-Terpinene

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Product Certifications

RUO – Research Use Only