missing translation for 'onlineSavingsMsg'
Learn More

Terpinen-4-ol, 97%, ACROS Organics™

Brand:  Acros Organics 360021000

Code : 29

Additional Details : CAS Number : 562-74-3 Weight : 0.15000kg

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10394352

  • £85.80 / 100g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

Terpinen-4-ol
562-74-3
97%
154.25
4-Carvomenthenol
97%
97%
C10H18O
MFCD00001562

Specifications

Terpinen-4-ol
562-74-3
Authentic
97%
C10H18O
MFCD00001562
10, 8996
Glass bottle
0.929
Solubility in water: very slightly soluble. Other solubilities: soluble in alcohol
Clear colorless to slightly yellow
100g
97%
0.9290g/mL
Liquid
97%
154.25
06, 55
4-Carvomenthenol
1.4760 to 1.4820
-1° to +1° (20°C, 589nm) (neat)
212.0°C
82°C
Videos
SDS