missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terfenadine EP Impurity J, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Terfenadine EP Impurity J
Specifications
Specifications
| Chemical Name or Material | Ethyl 1-[(4RS)-4-[4-(1,1-Dimethylethyl)phenyl]-4-hydroxybutyl]-piperidine-4-carboxylate |
| CAS | 100499-85-2 |
| Analyte or Component Names | Ethyl 1-[(4RS)-4-[4-(1,1-Dimethylethyl)phenyl]-4-hydroxybutyl]-piperidine-4-carboxylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H35 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ethyl 1-[(4RS)-4-[4-(1,1-Dimethylethyl)phenyl]-4-hydroxybutyl]-piperidine-4-carboxylate, Terfenadine Imp. J (EP), Terfenadine Impurity J |
| InChI Formula | InChI=1S/C22H35NO3/c1-5-26-21(25)18-12-15-23(16-13-18)14-6-7-20(24)17-8-10-19(11-9-17)22(2,3)4/h8-11,18,20,24H,5-7,12-16H2,1-4H3 |
| SMILES | CCOC(=O)C1CCN(CCCC(O)c2ccc(cc2)C(C)(C)C)CC1 |
| IUPAC Name | ethyl 1-[4-(4-tert-butylphenyl)-4-hydroxybutyl]piperidine-4-carboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction