missing translation for 'onlineSavingsMsg'
Learn More

Terephthaloyl Chloride, 99+%, ACROS Organics™

Brand:  Acros Organics 153102500

Code : 29

Additional Details : CAS Number : 100-20-9 Weight : 0.25000kg Transport : UN number : 2923 Chem class : 1a Pack group : I


Quantity 250g
Packaging Glass bottle

 View more versions of this product

Product Code. 10754091

  • £24.70 / 250g


EU Stock 6
Add to basket


Specifications

Chemical Identifiers

Terephthaloyl chloride
100-20-9
99+%
C6H4(COCl)2
MFCD00000693
99+%
≥99.0%
C8H4Cl2O2
203.02
1, 4-Benzenedicarbonyl chloride, TCl

Specifications

Terephthaloyl chloride
100-20-9
Crystalline Flakes, Crystals, and or Chunks
99+%
C6H4(COCl)2
MFCD00000693
1, 4-Benzenedicarbonyl chloride, TCl
White
250g
99+%
Authentic
≥99.0%
C8H4Cl2O2
203.02
09, 844
Glass bottle
79° to 83°C
Videos
SDS