missing translation for 'onlineSavingsMsg'
Learn More

Terephthaloyl Chloride, 99+%, ACROS Organics™

Brand:  Acros Organics 153100010

Code : 29

Additional Details : CAS Number : 100-20-9 Weight : 1.00000kg Transport : UN number : 2923 Chem class : 1a Pack group : I


Quantity 1kg
Packaging Glass bottle

 View more versions of this product

Product Code. 10307330

  • £68.60 / 1kg


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

Terephthaloyl chloride
100-20-9
99+%
C6H4(COCl)2
MFCD00000693
99+%
≥99.0%
C8H4Cl2O2
203.02
1, 4-Benzenedicarbonyl chloride

Specifications

Terephthaloyl chloride
100-20-9
Crystalline Flakes, Crystals, and or Chunks
99+%
C6H4(COCl)2
MFCD00000693
1, 4-Benzenedicarbonyl chloride
White
1kg
99+%
Authentic
≥99.0%
C8H4Cl2O2
203.02
09, 844
Glass bottle
79° to 83°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Terephthaloyl chloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS