missing translation for 'onlineSavingsMsg'
Learn More

Terephthalic acid, 99+%, Thermo Scientific Chemicals

Catalog Number 13458418
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
100-21-0
C8H6O4
166.13
MFCD00002558
KKEYFWRCBNTPAC-UHFFFAOYSA-N
p-phthalic acid, 1,4-benzenedicarboxylic acid, benzene-1,4-dicarboxylic acid, p-dicarboxybenzene, p-benzenedicarboxylic acid, p-carboxybenzoic acid, acide terephtalique, para-phthalic acid, tephthol, 1,4-dicarboxybenzene
7489
CHEBI:15702
terephthalic acid
C1=CC(=CC=C1C(=O)O)C(=O)O
100-21-0
C8H6O4
166.13
MFCD00002558
KKEYFWRCBNTPAC-UHFFFAOYSA-N
p-phthalic acid, 1,4-benzenedicarboxylic acid, benzene-1,4-dicarboxylic acid, p-dicarboxybenzene, p-benzenedicarboxylic acid, p-carboxybenzoic acid, acide terephtalique, para-phthalic acid, tephthol, 1,4-dicarboxybenzene
7489
CHEBI:15702
terephthalic acid
C1=CC(=CC=C1C(=O)O)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Fibre drum
Quantity 25 kg
Beilstein 09,841
Merck Index 15,9305
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkalies, slightly soluble in alcohol, insoluble in chloroform, ether and acetic acid
Sublimation Point 0402.0°C
Formula Weight 166.13
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Terephthalic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'