Learn More
Alfa Aesar™ Terbium vanadium oxide, 99.8% (REO)
Brand: Alfa Aesar™ 041995.09
Code : NEW
Additional Details : CAS Number : 13566-09-1 Weight : 0.01000kg Transport : UN number : 3288 Chem class : 1 Pack group : III
Quantity | 10g |
---|
Chemical Identifiers
Terbium vanadium oxide | |
273.87 | |
UN3288 |
13566-09-1 | |
MFCD00467967 | |
Terbium vanadate |
Specifications
Terbium vanadium oxide | |
Powder | |
(REO) | |
MFCD00467967 | |
Terbium vanadate |
13566-09-1 | |
99.8% | |
273.87 | |
UN3288 | |
10g |