missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Terbium(III,IV) oxide, 99.9% (REO)

Brand:  Alfa Aesar™ 036246.09

Code : NEW

Additional Details : CAS Number : 12037-01-3 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 11323308

  • £495.44 / 10g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

Terbium(III,IV) oxide
747.69
14,9157
12037-01-3
MFCD00083151

Specifications

Terbium(III,IV) oxide
Sintered Lump
(REO)
MFCD00083151
Odorless
2340°C
12037-01-3
99.9%
747.69
14,9157
3 to 12mm
10g
Videos
SDS