missing translation for 'onlineSavingsMsg'
Learn More

Terbium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11311799
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
7440-27-9
Tb
158.93
MFCD00011256
GZCRRIHWUXGPOV-UHFFFAOYSA-N
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
23958
CHEBI:33376
terbium
[Tb]
This item is not returnable. View return policy
7440-27-9
Tb
158.93
MFCD00011256
GZCRRIHWUXGPOV-UHFFFAOYSA-N
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
23958
CHEBI:33376
terbium
[Tb]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Quantity 25 x 25 mm
Percent Purity 99.9%
Packaging Mylar under Argon
Weight ≈1.29g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Terbium foil, 0.25mm (0.01 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'