missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Terbium foil, 0.127mm (0.005 in.) thick, 99.9% (REO)

Brand:  Alfa Aesar™ 000321.FF

Code : NEW

Additional Details : CAS Number : 7440-27-9 Weight : 0.01000kg

Quantity 25 x 25mm

 View more versions of this product

Product Code. 11316527

  • £50.30 / 25mm


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

Terbium foil
7440-27-9
0.127mm (0.005 in.) thick
MFCD00011256

Specifications

Terbium foil
25 x 25mm
7440-27-9
Foil
(REO)
≈0.66g/25x25mm
0.127mm (0.005 in.) thick
Mylar under Argon
99.9%
MFCD00011256
Videos
SDS