missing translation for 'onlineSavingsMsg'
Learn More

Terbium Foil, 0.025mm (0.001 in.) thick, (REO), 0.999, Alfa Aesar™

Form: Approx. 0.13g/25 x 25mm

Brand:  Alfa Aesar 012431.FF

Code : NEW

Additional Details : CAS Number : 7440-27-9 Weight : 0.01000kg Transport : UN number : 3178 Chem class : 1 Pack group : II

Quantity 25 x 25mm

 View more versions of this product

Product Code. 11331799

  • £161.12 / 25mm


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Terbium
Foil, 0.025mm (0.001 in.) thick
MFCD00011256
7440-27-9
99.9% (REO)

Specifications

Terbium
Foil, 0.025mm (0.001 in.) thick
Silver Gray foil
Silver-gray
7440-27-9
99.9% (REO)
MFCD00011256
25 x 25mm
Videos
SDS