missing translation for 'onlineSavingsMsg'
Learn More

TER-119 Monoclonal Antibody (TER-119), PE-Cyanine7, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

£94.75 - £149.00

Specifications

Antigen TER-119
Clone TER-119
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15330990
View Documents Promotion Details
Affymetrix eBioscience
25-5921-81
50 μg
£94.75
50µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15340990
View Documents Promotion Details
Affymetrix eBioscience
25-5921-82
100 μg
£149.00
100µg
View alternative products
Save Up to 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: The TER-119 monoclonal antibody reacts with mouse erythroid cells from early proerythroblast to mature erythrocyte stages. The TER-119 antigen is present in yolk sac, fetal and newborn liver, but is not expressed by cells carrying BFU-E and CFU-E activities. Several erythroleukemia cell lines tested so far are negative for expression of TER-119 antigen even after dimethylsulfoxide stimulation. Biochemical and molecular analysis of the TER-119 antigen indicate that this molecule is associated with the surface glycophorin A, but is not a typical glycophorin. Applications Reported: This TER-119 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TER-119 antibody has been tested by flow cytometric analysis of mouse splenocytes and bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat.

TER-119 Monoclonal antibody specifically detects TER-119 in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

TER-119
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
Ly76; Ly-76; TER119
Ly76
Primary
4° C, store in dark, DO NOT FREEZE!
Ly76
TER-119
Flow Cytometry
PE-Cyanine7
Rat
Mouse
0
104231
IgG2b κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.