missing translation for 'onlineSavingsMsg'
Learn More

TER-119, eFluor 450, clone: TER-119, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-5921-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15371800

  • £86.02 / 25µg


EU Stock 2
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The TER-119 monoclonal antibody reacts with mouse erythroid cells from early proerythroblast to mature erythrocyte stages. The TER-119 antigen is present in yolk sac, fetal and newborn liver, but is not expressed by cells carrying BFU-E and CFU-E activities. Several erythroleukemia cell lines tested so far are negative for expression of TER-119 antigen even after dimethylsulfoxide stimulation. Biochemical and molecular analysis of the TER-119 antigen indicate that this molecule is associated with the surface glycophorin A, but is not a typical glycophorin.Applications Reported: This TER-119 antibody has been reported for use in flow cytometric analysis.Applications Tested: This TER-119 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

TER-119
TER-119
eFluor 450
Conjugated
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TER119, Erythroid cell marker, Ly-76, Ly76
IgG2b
RUO
Primary
Murine
Videos