Learn More
TER-119 Monoclonal Antibody (TER-119), eFluor™ 450, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£109.00 - £219.00
Specifications
Antigen | TER-119 |
---|---|
Clone | TER-119 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15371800
|
Affymetrix eBioscience
48-5921-80 |
25 μg |
£109.00
25µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15381800
|
Affymetrix eBioscience
48-5921-82 |
100 μg |
£219.00
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The TER-119 monoclonal antibody reacts with mouse erythroid cells from early proerythroblast to mature erythrocyte stages. The TER-119 antigen is present in yolk sac, fetal and newborn liver, but is not expressed by cells carrying BFU-E and CFU-E activities. Several erythroleukemia cell lines tested so far are negative for expression of TER-119 antigen even after dimethylsulfoxide stimulation. Biochemical and molecular analysis of the TER-119 antigen indicate that this molecule is associated with the surface glycophorin A, but is not a typical glycophorin. Applications Reported: This TER-119 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TER-119 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 450 is an alternative to Pacific Blue™. eFluor™ 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.
TER-119 Monoclonal antibody specifically detects TER-119 in Mouse samples. It is validated for Flow Cytometry.Specifications
TER-119 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Ly76; Ly-76; TER119 | |
Ly76 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Ly76 |
TER-119 | |
Flow Cytometry | |
eFluor 450 | |
Rat | |
Mouse | |
0 | |
104231 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.