missing translation for 'onlineSavingsMsg'
Learn More

TER-119, clone: TER-119, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-5921-85

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15287597

  • £146.13 / 500µg


EU Stock 2
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The TER-119 monoclonal antibody reacts with mouse erythroid cells from early proerythroblast to mature erythrocyte stages. The TER-119 antigen is present in yolk sac, fetal and newborn liver, but is not expressed by cells carrying BFU-E and CFU-E activities. Several erythroleukemia cell lines tested so far are negative for expression of TER-119 antigen even after dimethylsulfoxide stimulation. Biochemical and molecular analysis of the TER-119 antigen indicate that this molecule is associated with the surface glycophorin A, but is not a typical glycophorin.Applications Reported: The TER-119 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunoblotting (WB), and immunohistochemical staining.Applications Tested: The TER-119 antibody has been tested by flow cytometric analysis of mouse splenocytes and bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

TER-119
TER-119
Unconjugated
Purified
Rat
500μg
Store at 2-8°C.
Monoclonal
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TER119, Erythroid cell marker, Ly-76, Ly76
IgG2b
RUO
Primary
Murine
Videos