missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TentaGel™ HL-S-Trityl, ACROS Organics™
Brand: Acros Organics 354470010
Code : 29
Additional Details : Weight : 0.05100kg
Quantity | 1g |
---|---|
Packaging | Glass bottle |
Description
For solid support synthesisChemical Identifiers
TentaGel™ HL-S-Trityl | |
MFCD01868633 | |
O-[2-(3-Tritylthiopropionamido)ethyl]polyethylene glycol polymer bound |
Specifications
TentaGel™ HL-S-Trityl | |
Glass bottle | |
1g | |
Powder | |
O-[2-(3-Tritylthiopropionamido)ethyl]polyethylene glycol polymer bound |
0.4 to 0.6mmol/g | |
mean particle size: 110 Micron | |
White to light yellow | |
MFCD01868633 |