Learn More
Tenovin-1, Tocris Bioscience™
Protects against MDM2-mediated p53 degradation
Brand:  Tocris (Bio-Techne) 336510
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13462894
  • £66.15 / 10mg
Estimated Shipment date
from Supplier 26-03-2020
Description
Description
p53 activator that protects against MDM2-mediated p53 degradation. Elevates levels of p53 and p21CIP/WAF1 and induces expression from an endogenous p53-dependent promoter. Exhibits potent antiproliferative activity in vitro.
Specifications
Chemical Identifiers
380315-80-0
369.483
tenovin-1, tenovin 1, unii-biq6aid2b7, biq6aid2b7, n-4-acetamidophenyl carbamothioyl-4-tert-butylbenzamide, n-4-acetamidophenyl carbamothioyl-4-tert-butyl benzamide, n-4-4-tert-butylphenyl formamido methanethioyl amino phenyl acetamide, dsstox_cid_26920, dsstox_rid_82016, dsstox_gsid_46920
N-[(4-acetamidophenyl)carbamothioyl]-4-tert-butylbenzamide
C20H23N3O2S
WOWJIWFCOPZFGV-UHFFFAOYSA-N
1013376
CC(=O)NC1=CC=C(C=C1)NC(=S)NC(=O)C2=CC=C(C=C2)C(C)(C)C
Specifications
10mg
380315-80-0
tenovin-1, tenovin 1, unii-biq6aid2b7, biq6aid2b7, n-4-acetamidophenyl carbamothioyl-4-tert-butylbenzamide, n-4-acetamidophenyl carbamothioyl-4-tert-butyl benzamide, n-4-4-tert-butylphenyl formamido methanethioyl amino phenyl acetamide, dsstox_cid_26920, dsstox_rid_82016, dsstox_gsid_46920
CC(=O)NC1=CC=C(C=C1)NC(=S)NC(=O)C2=CC=C(C=C2)C(C)(C)C
369.483
369.48
Tenovin-1
C20H23N3O2S
WOWJIWFCOPZFGV-UHFFFAOYSA-N
N-[(4-acetamidophenyl)carbamothioyl]-4-tert-butylbenzamide
1013376
>99%