Learn More
Tocris Bioscience™ Tenofovir
Reverse transcriptase inhibitor
Brand:  Tocris Bioscience™ 366650
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13443384
  • £555.12 / 50mg
Estimated Shipment
09-07-2019
Description
Description
Selectively inhibits HIV reverse transcriptase (RNA-dependent DNA polymerase). Prevents cytotoxicity in SIV-infected C-8166 cells in vitro (IC50 = 1.5 μM). Antiviral agent.
Specifications
Specifications
Tenofovir
>99
50mg
Tenofovir
287.21