missing translation for 'onlineSavingsMsg'
Learn More

LCR Hallcrest™ Temperature Strip

Brand:  LCR Hallcrest™ A296-04

Code : 03

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 13039232

  • £27.92 / Pack of 10


Estimated Shipment
20-06-2018
Add to basket

Specifications

Specifications

5°C
60 to 90°C
0.02
Videos