Learn More
BINDER™ Pt 100 Temperature Sensor
BINDER™ Pt 100 Temperature Sensor is an additional flexible Pt 100 for use with BD 23 and BD 400 Standard Incubators.
Brand: BINDER™ 8012-0115
Additional Details : Weight : 1.50000kg
Description
- Additional flexible Pt 100, interior, with external connection incl. LEMO connector (3-pin)
Order Info
Includes
Specifications
Pt 100 temperature sensor - additional flexible Pt 100, interior, with external connection incl. LEMO connector (3-pin) | |
BINDER Chambers BD 23, BD 400 | |
Temperature Sensor | |
LEMO Connector (3-Pin) |