Learn More
Memmert™ Temperature Accessories for Oven
A range of cooled incubators incorporating Peltier technology
Brand:  Memmert™ HPP110V3
Code : 50
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12667047
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
A range of cooled incubators constructed from rugged, hygienic, stainless steel material (430) incorporating Peltier technology with adaptive, fuzzy supported microprocessor PID controller. For stepless control of the Peltier heating/cooling system without refrigerant. Two PT-100 sensors Class A in a 4 wire circuit for function transfer, plus a seven day programme timer and ramp timer for up to 40 ramps, each adjustable from 1min to 999hr. Resolution of display for setpoint, actual values and over-temperature 0.1°C. Multiple over-temperature protection with audible and visual alarm: digital electronic over-temperature controller Class 3.3 for min. /under- and max. /over-temperature, Automatic Safety Function for over and under-temperature. Long term documentation through ring-store as data logger over six months - 1,024kB. A parallel printer interface for PCL3 compatible ink jet printers plus RS232 alternatively RS485 including software Celsius 2005.Memory card for 40 ramps 32kB and works calibration certificate for 10°C and 37°C.
Specifications
Specifications
Models 30, 55, 75, 110, 160, 260, 450, 750, 1060
-10°C to +80°C
Loop Interface
4 to 20 mA