Learn More
Temozolomide, Tocris Bioscience™
DNA-methylating antitumor agent
Brand:  Tocris (Bio-Techne) 270650
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13441764
  • £324.73 / 50mg
Estimated Shipment
27-06-2019
Description
Description
DNA methylating, chemotherapeutic agent. Displays antitumor activity against a board spectrum of tumors, including leukemias, lymphomas and solid tumors (IC50 = 5.0 μM for cytotoxicity against mouse TLX5 lymphoma cells). Induces autophagy in malignant glioma cells. Exhibits enhanced activity when used in combination with CHC (Cat No. 5029).
Specifications
Chemical Identifiers
85622-93-1
194.154
temozolomide, methazolastone, temodal, temodar, temozolamide, temozolomidum, 3-methyl-4-oxo-3,4-dihydroimidazo 5,1-d 1,2,3,5 tetrazine-8-carboxamide, temozolomidum latin, temozolodida spanish, temozolodida
CHEBI:72564
CN1C(=O)N2C=NC(=C2N=N1)C(=O)N
C6H6N6O2
BPEGJWRSRHCHSN-UHFFFAOYSA-N
5394
3-methyl-4-oxoimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxamide
Specifications
Temozolomide
85622-93-1
temozolomide, methazolastone, temodal, temodar, temozolamide, temozolomidum, 3-methyl-4-oxo-3,4-dihydroimidazo 5,1-d 1,2,3,5 tetrazine-8-carboxamide, temozolomidum latin, temozolodida spanish, temozolodida
CN1C(=O)N2C=NC(=C2N=N1)C(=O)N
194.154
CHEBI:72564
>99%
50mg
C6H6N6O2
BPEGJWRSRHCHSN-UHFFFAOYSA-N
3-methyl-4-oxoimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxamide
5394
194.15
Videos