missing translation for 'onlineSavingsMsg'
Learn More

Tellurium Standard For ICP, 1000μg/mL in 5% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep PLTE2-2Y

Code : 04

Additional Details : CAS Number : 13494-80-9 Weight : 0.12800kg Transport : UN number : 3264 Chem class : 1a Pack group : III

Quantity 125mL
Packaging Triple-leached LDPE bottle

 View more versions of this product

Product Code. 10056523

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

AnalyteConcentration
Te1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

Tellurium Standard
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Specifications

Tellurium Standard
94.9
Assurance Grade
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
125mL
7732-18-5
94.9
Matrix&tab;Concentration&tab;Cat. No.
2-5% HNO3&tab;1000μg/mL&tab;PLTE2-2Y, -2X
Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
5% HNO3
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Videos
Documentation

Product intended for laboratory use only