missing translation for 'onlineSavingsMsg'
Learn More

Tellurium powder, -30 mesh, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048635
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
13494-80-9
Te
127.60
MFCD00134062
PORWMNRCUJJQNO-UHFFFAOYSA-N
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
6327182
CHEBI:30452
tellurium
[Te]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
13494-80-9
Te
127.60
MFCD00134062
PORWMNRCUJJQNO-UHFFFAOYSA-N
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
6327182
CHEBI:30452
tellurium
[Te]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
UN Number UN3288
Quantity 25 g
Percent Purity 99.99%
Mesh Size −30 Mesh
Odor Odorless
Form Powder
Assay (metals basis)
Chemical Name or Material Tellurium powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'