missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tellurium(IV) isopropoxide, 99.9% (metals basis)

Brand:  Alfa Aesar™ 036565.09

Code : NEW

Additional Details : CAS Number : 1795-64-8 Weight : 0.01000kg Transport : UN number : 1993 Chem class : 1 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 11307998

  • £477.32 / 10g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Chemical Identifiers

Tellurium(IV) isopropoxide
363.73
UN1993
1795-64-8
MFCD00145354

Specifications

Tellurium(IV) isopropoxide
Liquid
(metals basis)
MFCD00145354
76°C (0.5mmHg)
10g
1795-64-8
99.9%
363.73
UN1993
Alcohol-like
Moisture & Light Sensitive
Videos
SDS