missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tellurium(IV) ethoxide, tech. 85%, remainder ethanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
Description
Tellurium(IV) ethoxide could be used in sol-gel processing for the fabrication of tellurite thin films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >100°C (decomposition) |
Density | 1.52 g/mL |
Boiling Point | 88°C to 90°C (2 mmHg) |
Flash Point | 27°C (81°F) |
Odor | Alcohol-like |
Quantity | 2 g |
UN Number | UN1993 |
Sensitivity | Light and moisture sensitive |
Synonym | tetraethoxytellurium iv, tellurium iv ethoxide, triethoxy-??-tellanyl oxy ethane |
Solubility Information | Decomposes in water to TeO2 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'