missing translation for 'onlineSavingsMsg'
Learn More

Tellurium(IV) ethoxide, tech. 85%, remainder ethanol, Thermo Scientific Chemicals

Catalog Number 11309527
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
2017-01-8
C6H14N2O
130.19
MFCD00270937
RWIVICVCHVMHMU-UHFFFAOYSA-N
408285
2-morpholin-4-ylethanamine
C1COCCN1CCN
This item is not returnable. View return policy
2017-01-8
C6H14N2O
130.19
MFCD00270937
RWIVICVCHVMHMU-UHFFFAOYSA-N
408285
2-morpholin-4-ylethanamine
C1COCCN1CCN

Tellurium(IV) ethoxide could be used in sol-gel processing for the fabrication of tellurite thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >100°C (decomposition)
Density 1.52 g/mL
Boiling Point 88°C to 90°C (2 mmHg)
Flash Point 27°C (81°F)
Odor Alcohol-like
Quantity 10 g
UN Number UN1993
Sensitivity Light and moisture sensitive
Synonym 4-2-aminoethyl morpholine, 2-morpholinoethanamine, 4-morpholineethanamine, 2-morpholinoethylamine, n-aminoethylmorpholine, 2-morpholin-4-yl ethan-1-amine, morpholine, 4-2-aminoethyl, n-2-aminoethyl morpholine, n-2-aminoethylmorpholine, 2-morpholin-4-ylethylamine
Solubility Information Decomposes in water to TeO2
Formula Weight 307.84
Percent Purity 85%
Grade Technical
Physical Form Liquid
Assay Remainder ethanol
Chemical Name or Material Tellurium(IV) ethoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'