missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Telluric acid dihydrate, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Telluric acid dihydrate is used in the preparation of tellurium nanorods through hydrazine-driven reduction, which finds application in solar cells, thermoelectric and topological insulators. It is also involved in the preparation of tellurium metal by reacting with the solution of potassium thiocyanate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 160°C (decomposition) |
Melting Point | 136°C |
Color | White |
Physical Form | Crystalline |
Quantity | 10 g |
Assay Percent Range | >99% |
Solubility Information | Soluble in water. |
Formula Weight | 229.64 (193.61 Anhydrous) |
Sensitivity | Hygroscopic |
Density | 3.158 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'