missing translation for 'onlineSavingsMsg'
Learn More

Telenzepine dihydrochloride, Tocris Bioscience™

Potent, selective M1 antagonist

Brand:  Tocris (Bio-Techne) 112250

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13438603

  • £266.40 / 50mg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective high affinity muscarinic M1 receptor antagonist (Ki = 0.94 nM). Inhibits gastric acid secretion.
Specifications

Chemical Identifiers

147416-96-4
443.387
telenzepine dihydrochloride, telenzepine hydrochloride, prestwick_649, 4,9-dihydro-3-methyl-4-4-methyl-1-piperazinyl acetyl-10h-thieno 3,4-b 1,5 benzodiazepin-10-one dihydrochloride, 4,9-dihydro-3-methyl-4-4-methyl-1, piperazinyl acetyl-10h-thieno 3,4-b 1,5 benzodiazepin-10-one dihydrochloride, 1-methyl-10-2-4-methylpiperazin-1-yl acetyl-5h-thieno 3,4-b 1,5 benzodiazepin-4-one dihydrochloride
1-methyl-10-[2-(4-methylpiperazin-1-yl)acetyl]-5H-thieno[3,4-b][1,5]benzodiazepin-4-one;dihydrochloride
C19H24Cl2N4O2S
JTQJFSQQHGPLOX-UHFFFAOYSA-N
6419995
CC1=C2C(=CS1)C(=O)NC3=CC=CC=C3N2C(=O)CN4CCN(CC4)C.Cl.Cl

Specifications

Telenzepine dihydrochloride
147416-96-4
telenzepine dihydrochloride, telenzepine hydrochloride, prestwick_649, 4,9-dihydro-3-methyl-4-4-methyl-1-piperazinyl acetyl-10h-thieno 3,4-b 1,5 benzodiazepin-10-one dihydrochloride, 4,9-dihydro-3-methyl-4-4-methyl-1, piperazinyl acetyl-10h-thieno 3,4-b 1,5 benzodiazepin-10-one dihydrochloride, 1-methyl-10-2-4-methylpiperazin-1-yl acetyl-5h-thieno 3,4-b 1,5 benzodiazepin-4-one dihydrochloride
CC1=C2C(=CS1)C(=O)NC3=CC=CC=C3N2C(=O)CN4CCN(CC4)C.Cl.Cl
443.387
443.39
50mg
C19H24Cl2N4O2S
JTQJFSQQHGPLOX-UHFFFAOYSA-N
1-methyl-10-[2-(4-methylpiperazin-1-yl)acetyl]-5H-thieno[3,4-b][1,5]benzodiazepin-4-one;dihydrochloride
6419995
>98%
Videos
Documents