missing translation for 'onlineSavingsMsg'
Learn More

Tegaserod maleate, Tocris Bioscience™

5-HT4 partial agonist

Brand:  RD Systems (Bio-Techne) 4613/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13464984

  • £135.00 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Partial agonist of 5-HT4 (Ki = 12 nM).
Specifications

Chemical Identifiers

189188-57-6
417.466
tegaserod maleate, sdz-htf 919, 3-5-methoxy-1h-indol-3-ylmethylene-n-pentylcarbazimidamide hydrogen maleate, 2-5-methoxy-1h-indol-3-yl methylene-n-pentylhydrazinecarboximidamide 2z-but-2-enedioic acid
(Z)-but-2-enedioic acid;1-[(5-methoxyindol-3-ylidene)methylamino]-2-pentylguanidine
C20H27N5O5
KIJRKSHEIAFGBH-BTJKTKAUSA-N
57149572
CCCCCN=C(N)NNC=C1C=NC2=C1C=C(C=C2)OC.C(=CC(=O)O)C(=O)O

Specifications

Tegaserod maleate
189188-57-6
tegaserod maleate, sdz-htf 919, 3-5-methoxy-1h-indol-3-ylmethylene-n-pentylcarbazimidamide hydrogen maleate, 2-5-methoxy-1h-indol-3-yl methylene-n-pentylhydrazinecarboximidamide 2z-but-2-enedioic acid
CCCCCN=C(N)NNC=C1C=NC2=C1C=C(C=C2)OC.C(=CC(=O)O)C(=O)O
417.466
417.46
10mg
C20H27N5O5
KIJRKSHEIAFGBH-BTJKTKAUSA-N
(Z)-but-2-enedioic acid;1-[(5-methoxyindol-3-ylidene)methylamino]-2-pentylguanidine
57149572
>99%
Videos