missing translation for 'onlineSavingsMsg'
Learn More

Technical Buffer Solution pH 7, WTW™

Catalog Number p-8014339
Click to view available options
Quantity:
250 mL
1000 mL
Packaging:
1 Bottle
10 Bottles
25 Bottles
This item is not returnable. View return policy
This item is not returnable. View return policy

Precise and traceable to NIST (National Institute of Standards and Technology), their accuracy is ± 0,03 pH.

  • Precise and traceable to NIST (National Institute of Standards and Technology, USA in bulk size of 250 ml with built-in dosing vessel standard buffer
  • Life up to 3 years (unopened)
  • Thrifty use
  • Easiest handling by means of dosing device
pH 7
Physical Form Liquid
Quantity 250 mL
Packaging 1 Bottle
Traceability to NIST Yes
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'