Learn More
Bibby Scientific™ Techne™ Elma Tec Cleaning Reagent
Brand: Bibby Scientific™ 8000141
Additional Details : Weight : 0.85000kg Transport : UN number : 3253 Chem class : 8 Pack group : III
Specifications
0.85Kg | |
Elma Tec Cleaning Reagent |
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : cocosfattyaminoxethylate, tatrasodium pyrophosphate, sodiumcarbinate, disodiummetasilicate, decan-